Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abl2
Entrez Gene ID
304883
Gene Name
ABL proto-oncogene 2, non-receptor tyrosine kinase
Gene Aliases
Abll, Arg
Chromosome Location
Chr.13: 74154533 - 74333244 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107186.1 NP_001100656.1 11 - 12 1822 118
XM_006250047.2 XP_006250109.1 11 - 12 2130 118
XM_006250049.2 XP_006250111.1 11 - 12 2800 118
XM_006250051.2 XP_006250113.1 10 - 11 2067 118
XM_006250053.2 XP_006250115.1 11 - 12 2129 118
XM_006250054.2 XP_006250116.1 11 - 12 2799 118
XM_006250055.2 XP_006250117.1 10 - 11 2066 118

View Full Product Details