Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc27a6
Entrez Gene ID
291582
Gene Name
solute carrier family 27 (fatty acid transporter), member 6
Gene Aliases
-
Chromosome Location
Chr.18: 53609497 - 53667833 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106145.1 NP_001099615.1 1 - 2 706 70

View Full Product Details