Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slco1b2
Entrez Gene ID
58978
Gene Name
solute carrier organic anion transporter family, member 1B2
Gene Aliases
OATP-4, Oatp4, Slc21a10, Slco1b3, rlst-1
Chromosome Location
Chr.4: 175814118 - 175881775 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270586.1 NP_001257515.1 7 - 8 834 156
NM_031650.3 NP_113838.1 7 - 8 834 156
GenBank mRNA AF147740.1 - 6 - 7 720 156
AF208545.2 - 7 - 8 834 156
AJ271682.1 - 7 - 8 835 156

View Full Product Details