Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mynn
Entrez Gene ID
361924
Gene Name
myoneurin
Gene Aliases
-
Chromosome Location
Chr.2: 116397154 - 116414788 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012178.1 NP_001012178.1 3 - 4 999 117
XM_006232232.2 XP_006232294.1 3 - 4 2043 117
XM_006232233.2 XP_006232295.1 3 - 4 2044 117
XM_006232234.2 XP_006232296.1 3 - 4 1913 117
GenBank mRNA BC089853.1 - 3 - 4 999 117

View Full Product Details