Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skil
Entrez Gene ID
114208
Gene Name
SKI-like proto-oncogene
Gene Aliases
Skir, Sno
Chromosome Location
Chr.2: 115862932 - 115891309 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232237.2 XP_006232299.1 4 - 5 2478 150
XM_008760948.1 XP_008759170.1 3 - 4 3484 150

View Full Product Details