Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arsk
Entrez Gene ID
365619
Gene Name
arylsulfatase family, member K
Gene Aliases
RGD1310182
Chromosome Location
Chr.2: 2835568 - 2891059 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047917.1 NP_001041382.1 6 - 7 1087 114

View Full Product Details