Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mocs2
Entrez Gene ID
294753
Gene Name
molybdenum cofactor synthesis 2
Gene Aliases
-
Chromosome Location
Chr.2: 46980976 - 46992886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007633.2 NP_001007634.1 5 - 6 676 107
NM_001162413.1 NP_001155885.1 6 - 7 798 107
XM_006231985.2 XP_006232047.1 6 - 7 744 107
XM_006231986.2 XP_006232048.1 5 - 6 641 107
GenBank mRNA BC079181.1 - 6 - 7 764 107
FQ220090.1 - 6 - 7 693 107
FQ220210.1 - 5 - 6 589 107
FQ230755.1 - 5 - 6 599 107
FQ235152.1 - 6 - 7 727 107

View Full Product Details