The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173126.3 | NP_775149.1 | 1 - 2 | 345 | 124 |
GenBank mRNA | AF313411.1 | - | 1 - 2 | 144 | 124 |
BC070515.1 | - | 1 - 2 | 345 | 124 | |
EV780394.1 | - | 1 - 2 | 126 | 124 |