Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serp1
Entrez Gene ID
80881
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4
Chromosome Location
Chr.2: 148718279 - 148722263 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030835.2 NP_110462.1 2 - 3 522 81
GenBank mRNA AB018546.1 - 2 - 3 407 81
AF100470.1 - 2 - 3 176 81
AJ238236.1 - 2 - 3 182 81
BC061854.1 - 2 - 3 522 81

View Full Product Details