Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlx4
Entrez Gene ID
303469
Gene Name
distal-less homeobox 4
Gene Aliases
-
Chromosome Location
Chr.10: 82958525 - 82963919 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107040.1 NP_001100510.1 1 - 2 287 73
GenBank mRNA HQ616894.1 - 1 - 2 309 73

View Full Product Details