Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsg2
Entrez Gene ID
497878
Gene Name
neuron specific gene family member 2
Gene Aliases
-
Chromosome Location
Chr.10: 15867885 - 15928169 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034152.1 NP_001029324.1 1 - 2 110 79
GenBank mRNA BC105916.1 - 1 - 2 110 79

View Full Product Details