Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aatk
Entrez Gene ID
690853
Gene Name
apoptosis-associated tyrosine kinase
Gene Aliases
-
Chromosome Location
Chr.10: 109184338 - 109224482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168703.1 NP_001162174.1 5 - 6 676 84
XM_006247926.2 XP_006247988.1 5 - 6 788 84
XM_006247927.2 XP_006247989.1 4 - 5 469 84
XM_006247928.2 XP_006247990.1 5 - 6 680 84
XM_008768484.1 XP_008766706.1 5 - 6 701 84

View Full Product Details