The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168703.1 | NP_001162174.1 | 6 - 7 | 768 | 85 |
XM_006247926.2 | XP_006247988.1 | 6 - 7 | 880 | 85 | |
XM_006247927.2 | XP_006247989.1 | 5 - 6 | 561 | 85 | |
XM_006247928.2 | XP_006247990.1 | 6 - 7 | 772 | 85 | |
XM_008768484.1 | XP_008766706.1 | 6 - 7 | 793 | 85 |