The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105783.1 | NP_001099253.1 | 2 - 3 | 442 | 89 |
XM_006246448.2 | XP_006246510.1 | 2 - 3 | 451 | 89 |