Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd79b
Entrez Gene ID
171055
Gene Name
Cd79b molecule, immunoglobulin-associated beta
Gene Aliases
B29, Igb
Chromosome Location
Chr.10: 94497445 - 94500591 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133533.2 NP_598217.1 1 - 2 132 104
GenBank mRNA BC088115.1 - 1 - 2 132 104
FQ226266.1 - 1 - 2 129 104
FQ227788.1 - 1 - 2 121 104
FQ230261.1 - 1 - 2 123 104
FQ232054.1 - 1 - 2 123 104
FQ232680.1 - 1 - 2 123 104
FQ232790.1 - 1 - 2 104
FQ233507.1 - 1 - 2 121 104
FQ234082.1 - 1 - 2 106 104
FQ234487.1 - 1 - 2 121 104

View Full Product Details