Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nxn
Entrez Gene ID
360577
Gene Name
nucleoredoxin
Gene Aliases
-
Chromosome Location
Chr.10: 64411710 - 64550147 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108285.1 NP_001101755.1 4 - 5 583 87
XM_006246907.1 XP_006246969.1 4 - 5 783 87

View Full Product Details