The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108285.1 | NP_001101755.1 | 5 - 6 | 697 | 59 |
XM_006246907.1 | XP_006246969.1 | 5 - 6 | 897 | 59 |