Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stat5b
Entrez Gene ID
25126
Gene Name
signal transducer and activator of transcription 5B
Gene Aliases
-
Chromosome Location
Chr.10: 88686207 - 88712313 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022380.1 NP_071775.1 - 1492 72
GenBank mRNA X97541.1 - - 1492 72

View Full Product Details