Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkar1a
Entrez Gene ID
25725
Gene Name
protein kinase, cAMP-dependent, regulatory, type I, alpha
Gene Aliases
RIIA
Chromosome Location
Chr.10: 97940705 - 97959199 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013181.1 NP_037313.1 5 - 6 594 130
XM_008768311.1 XP_008766533.1 5 - 6 650 130
GenBank mRNA M17086.1 - 5 - 6 594 130
U75932.1 - 2 - 3 108 130

View Full Product Details