Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uprt
Entrez Gene ID
317237
Gene Name
uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae)
Gene Aliases
RGD1564806
Chromosome Location
Chr.X: 75382567 - 75408539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257154.2 XP_006257216.1 6 - 7 953 124
XM_228538.8 XP_228538.3 7 - 8 905 124

View Full Product Details