The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009953.2 | NP_001009953.2 | 6 - 7 | 792 | 87 |
NM_080909.2 | NP_543185.2 | 7 - 8 | 2966 | 87 | |
XM_006230649.2 | XP_006230711.1 | 8 - 9 | 3038 | 87 | |
XM_006230650.2 | XP_006230712.1 | 6 - 7 | 762 | 87 | |
GenBank mRNA | AF442357.1 | - | 6 - 7 | 796 | 87 |
AF442358.1 | - | 6 - 7 | 796 | 87 | |
AY164698.1 | - | 6 - 7 | 796 | 87 | |
AY496443.1 | - | 7 - 8 | 2966 | 87 | |
BC062068.1 | - | 6 - 7 | 790 | 87 | |
BC107448.1 | - | 6 - 7 | 792 | 87 | |
BC129077.1 | - | 6 - 7 | 800 | 88 |