Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phldb3
Entrez Gene ID
308431
Gene Name
pleckstrin homology-like domain, family B, member 3
Gene Aliases
RGD1561121
Chromosome Location
Chr.1: 81474553 - 81492631 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191622.1 NP_001178551.1 8 - 9 1160 128

View Full Product Details