Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ifnar2
Entrez Gene ID
686326
Gene Name
interferon (alpha, beta and omega) receptor 2
Gene Aliases
-
Chromosome Location
Chr.11: 31527615 - 31559834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248107.2 XP_006248169.1 8 - 9 1135 104

View Full Product Details