Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sntg2
Entrez Gene ID
298936
Gene Name
syntrophin, gamma 2
Gene Aliases
-
Chromosome Location
Chr.6: 49123084 - 49324530 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106720.1 NP_001100190.1 12 - 13 1372 100
XM_006239967.2 XP_006240029.1 16 - 17 1653 100
XM_006239968.2 XP_006240030.1 15 - 16 1533 100

View Full Product Details