The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013942.1 | NP_001013964.1 | 14 - 15 | 1910 | 148 |
GenBank mRNA | BC081910.1 | - | 14 - 15 | 1910 | 148 |