Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfc2
Entrez Gene ID
116468
Gene Name
replication factor C (activator 1) 2
Gene Aliases
-
Chromosome Location
Chr.12: 25130375 - 25143480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053786.1 NP_446238.1 3 - 4 246 90
GenBank mRNA BC082110.1 - 3 - 4 246 90

View Full Product Details