Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc42b
Entrez Gene ID
304509
Gene Name
coiled-coil domain containing 42B
Gene Aliases
RGD1561303
Chromosome Location
Chr.12: 41463694 - 41469917 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_222198.7 XP_222198.5 1 - 2 283 107

View Full Product Details