Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tspan33
Entrez Gene ID
500065
Gene Name
tetraspanin 33
Gene Aliases
RGD1560915
Chromosome Location
Chr.4: 56981283 - 57005866 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109227.1 NP_001102697.1 2 - 3 363 80

View Full Product Details