The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022522.2 | NP_071967.2 | 9 - 10 | 1128 | 150 |
XM_006236394.2 | XP_006236456.1 | 8 - 9 | 1809 | 150 | |
GenBank mRNA | AF136231.1 | - | 9 - 10 | 1126 | 150 |
AF136232.1 | - | 10 - 11 | 1187 | 150 | |
U77933.1 | - | 9 - 10 | 1128 | 150 |