Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wasl
Entrez Gene ID
682507
Gene Name
Wiskott-Aldrich syndrome-like
Gene Aliases
N-WASP, TRS4
Chromosome Location
Chr.4: 51677722 - 51726212 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110365.1 NP_001103835.1 2 - 3 614 161
GenBank mRNA D88461.1 - 2 - 3 320 161

View Full Product Details