Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lipm
Entrez Gene ID
309528
Gene Name
lipase, family member M
Gene Aliases
Lipl3
Chromosome Location
Chr.1: 252409232 - 252428567 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_220066.7 XP_220066.4 5 - 6 925 156

View Full Product Details