Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sigirr
Entrez Gene ID
309106
Gene Name
single immunoglobulin and toll-interleukin 1 receptor (TIR) domain
Gene Aliases
RGD1306732
Chromosome Location
Chr.1: 213898393 - 213907205 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024887.1 NP_001020058.1 6 - 7 823 137
XM_006230552.2 XP_006230614.1 6 - 7 1657 137
XM_006230553.2 XP_006230615.1 6 - 7 1289 137
XM_008759988.1 XP_008758210.1 5 - 6 2855 137
XM_008759989.1 XP_008758211.1 6 - 7 1348 137
XM_008759990.1 XP_008758212.1 6 - 7 851 137
GenBank mRNA BC097488.1 - 6 - 7 823 137

View Full Product Details