Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpm
Entrez Gene ID
314855
Gene Name
carboxypeptidase M
Gene Aliases
-
Chromosome Location
Chr.7: 60654350 - 60714620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108098.1 NP_001101568.1 6 - 7 814 73

View Full Product Details