Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem19
Entrez Gene ID
299800
Gene Name
transmembrane protein 19
Gene Aliases
-
Chromosome Location
Chr.7: 58316583 - 58343760 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199098.1 NP_954529.1 2 - 3 526 103
XM_006241347.2 XP_006241409.1 3 - 4 623 103
XM_006241348.2 XP_006241410.1 3 - 4 476 103
XM_006241349.1 XP_006241411.1 3 - 4 361 103
XM_008765355.1 XP_008763577.1 2 - 3 572 103
GenBank mRNA BC061871.1 - 2 - 3 526 103

Gene Symbol
LOC100912615
Entrez Gene ID
100912615
Gene Name
transmembrane protein 19-like
Gene Aliases
-
Chromosome Location
Chr.7: 57787695 - 57817537 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765371.1 XP_008763593.1 - 631 103

View Full Product Details