Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc16a7
- Entrez Gene ID
-
29735
- Gene Name
- solute carrier family 16 (monocarboxylate transporter), member 7
- Gene Aliases
-
Mct2
- Chromosome Location
-
Chr.7: 68445819 - 68606295 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U62316.1 |
- |
2 - 3 |
200 |
100 |
View Full Product Details