Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudcd1
Entrez Gene ID
362906
Gene Name
NudC domain containing 1
Gene Aliases
RGD1310624
Chromosome Location
Chr.7: 83302778 - 83348529 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130561.2 NP_001124033.1 5 - 6 873 149
GenBank mRNA BC169014.1 - 5 - 6 873 149

View Full Product Details