Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gli1
Entrez Gene ID
140589
Gene Name
GLI family zinc finger 1
Gene Aliases
Gli
Chromosome Location
Chr.7: 70620794 - 70633163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191910.1 NP_001178839.1 9 - 10 1306 80
XM_006241442.2 XP_006241504.1 11 - 12 1571 80
XM_006241443.2 XP_006241505.1 10 - 11 1454 80
XM_008765375.1 XP_008763597.1 11 - 12 1519 80
GenBank mRNA AB073717.1 - 1 - 2 239 80

View Full Product Details