Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chadl
Entrez Gene ID
100910434
Gene Name
chondroadherin-like
Gene Aliases
-
Chromosome Location
Chr.7: 122915847 - 122929114 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765808.1 XP_008764030.1 4 - 5 2356 84

View Full Product Details