Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Deptor
Entrez Gene ID
314979
Gene Name
DEP domain containing MTOR-interacting protein
Gene Aliases
Depdc6, RGD1561030
Chromosome Location
Chr.7: 94795159 - 95000750 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765525.1 XP_008763747.1 4 - 5 791 120

View Full Product Details