Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mterfd1
Entrez Gene ID
299514
Gene Name
MTERF domain containing 1
Gene Aliases
Cgi12, mTERF3
Chromosome Location
Chr.7: 71275544 - 71293811 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199387.1 NP_955419.1 4 - 5 778 150
XM_006241509.2 XP_006241571.1 4 - 5 804 150
GenBank mRNA BC062080.1 - 4 - 5 778 150

View Full Product Details