Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd17
Entrez Gene ID
300317
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
RGD1311154
Chromosome Location
Chr.7: 119647229 - 119677405 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134529.1 NP_001128001.1 5 - 6 683 96
XM_006241976.2 XP_006242038.1 5 - 6 829 96
XM_006241977.2 XP_006242039.1 5 - 6 823 96
XM_008765708.1 XP_008763930.1 5 - 6 826 96
XM_008765709.1 XP_008763931.1 5 - 6 824 96
XM_008765710.1 XP_008763932.1 5 - 6 827 96

View Full Product Details