Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd17
Entrez Gene ID
300317
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
RGD1311154
Chromosome Location
Chr.7: 119647229 - 119677405 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241983.2 XP_006242045.1 5 - 6 815 94

View Full Product Details