The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031335.3 | NP_112625.1 | 4 - 5 | 338 | 134 |
GenBank mRNA | AB017711.1 | - | 4 - 5 | 338 | 134 |
BC083552.1 | - | 4 - 5 | 378 | 134 |