Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rad21
Entrez Gene ID
314949
Gene Name
RAD21 homolog (S. pombe)
Gene Aliases
-
Chromosome Location
Chr.7: 91511755 - 91538673 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025701.1 NP_001020872.1 3 - 4 480 85
GenBank mRNA BC099200.1 - 3 - 4 480 85

View Full Product Details