Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rad21
Entrez Gene ID
314949
Gene Name
RAD21 homolog (S. pombe)
Gene Aliases
-
Chromosome Location
Chr.7: 91511755 - 91538673 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025701.1 NP_001020872.1 5 - 6 688 80
GenBank mRNA BC099200.1 - 5 - 6 688 80

View Full Product Details