The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130500.1 | NP_001123972.1 | 8 - 9 | 1740 | 71 |
XM_006242160.2 | XP_006242222.1 | 9 - 10 | 1844 | 71 | |
GenBank mRNA | FQ218328.1 | - | 9 - 10 | 1812 | 71 |