Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St3gal1
Entrez Gene ID
362924
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 1
Gene Aliases
Siat4a
Chromosome Location
Chr.7: 107895045 - 107963150 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013219.1 NP_001013237.1 2 - 3 500 75
XM_006241709.2 XP_006241771.1 5 - 6 1472 75
XM_006241710.2 XP_006241772.1 5 - 6 2390 75
XM_008765513.1 XP_008763735.1 4 - 5 1417 75
GenBank mRNA AJ748840.1 - 2 - 3 500 75

View Full Product Details