The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008378.1 | NP_001008379.1 | 5 - 6 | 652 | 61 |
GenBank mRNA | BC086452.1 | - | 5 - 6 | 652 | 61 |
FQ214182.1 | - | 5 - 6 | 622 | 61 | |
FQ219469.1 | - | 5 - 6 | 645 | 61 | |
FQ219661.1 | - | 5 - 6 | 650 | 61 |