Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom2r43
Entrez Gene ID
679593
Gene Name
vomeronasal 2 receptor, 43
Gene Aliases
-
Chromosome Location
Chr.1: 149506822 - 149529350 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099512.1 NP_001092982.1 2 - 3 480 140

View Full Product Details