Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsd3b7
Entrez Gene ID
246211
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Gene Aliases
Cca2
Chromosome Location
Chr.1: 199248084 - 199251747 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230212.2 XP_006230274.1 5 - 6 1029 158

View Full Product Details